accessibility menu, dialog, popup

UserName

H3CNCH3NH3CCH3ONClNNFPFFFFF

CAS RN 330645-87-9

IUPAC Name: hexafluoro-λ⁵-phosphanuide; {[(6-chloro-1H-1,2,3-benzotriazol-1-yl)oxy](dimethylamino)methylidene}dimethylazanium
Synonyms: {[(6-chloro-1,2,3-benzotriazol-1-yl)oxy](dimethylamino)methylidene}dimethylazanium hexafluorophosphate
Molecular Weight (g/mol): 413.69
Molecular Formula: C11H15ClF6N5OP
InChi Key: ZHHGTMQHUWDEJF-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=C(Cl)C=C12)=[N+](C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results