missing translation for 'onlineSavingsMsg'
Learn More

O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%

Product Code. 15572293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15572293 10 g 10g
15582293 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15572293 Supplier Thermo Scientific Acros Supplier No. 448930100
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 330645-87-9 | C11H15ClN5O·F6P | 413.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330645-87-9
Molecular Formula C11H15ClN5O·F6P
Molecular Weight (g/mol) 413.69
InChI Key ZHHGTMQHUWDEJF-UHFFFAOYSA-N
Synonym hctu, o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, 2-6-chloro-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate v, 1-bis-dimethylamino-methylene-5-chloro-3-oxy-1h-benzotriazol-1-ium hexafluorophosphate, 2-6-chloro-1h-benzotriazole-1-yl-1,1,3,3-tetramethylaminium hexafluorophosphate, o-1h-6-chlorobenzotriazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-6-chloro-1-hydrocibenzotriazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, 2-6-chloro-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethylisouronium hexafluorophosphate v, pubchem12740, acmc-20aj2u
PubChem CID 42624899
IUPAC Name [(6-chlorobenzotriazol-1-yl)oxy-(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)ON1C2=C(C=CC(=C2)Cl)N=N1.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 185.0°C to 190.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 10 g
Formula Weight 413.69
Percent Purity 98%
Chemical Name or Material O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate
compliance-icons
Product Identifier
  • O-(6-Chloro-1-hydrocibenzotriazol-1-yl)-1,1,3,3-tetramethyluronium hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.