accessibility menu, dialog, popup

UserName

CH3CH3H3COONHBOH3CH3CH3CCH3O

CAS RN 330793-09-4

IUPAC Name: tert-butyl N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Synonyms: tert-butyl N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Molecular Weight (g/mol): 319.21
Molecular Formula: C17H26BNO4
InChi Key: ANQAOGOIWVMGCH-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results