CAS RN 331862-41-0
CAS RN 331862-41-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
IMR-1A, MedChemExpress
MedChemExpress IMR-1A, a acid metabolite of IMR-1, is a Notch inhibitor with an IC50 of 0.5 μM. IMR-1A has a 50-fold increase in potency with respect to IMR-1. IMR-1 can metabolize in vivo to IMR-1A.