accessibility menu, dialog, popup

UserName

CAS RN 332117-28-9

IUPAC Name: N-[4-(butan-2-yl)phenyl]-2-(1,3-dimethyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-7-yl)acetamide
Synonyms: 2-(1,3-dimethyl-2,6-dioxopurin-7-yl)-N-[4-(sec-butyl)phenyl]acetamide
Molecular Weight (g/mol): 369.43
Molecular Formula: C19H23N5O3
InChi Key: ZUTUWJYMCADJHD-UHFFFAOYNA-N
SMILES: CCC(C)C1=CC=C(NC(=O)CN2C=NC3=C2C(=O)N(C)C(=O)N3C)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)