missing translation for 'onlineSavingsMsg'
Learn More

Chembridge-5861528, MedChemExpress

Product Code. 16496298
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496298 10 mg 10mg
16488887 5 mg 5mg
16482417 50 mg 50mg
16476626 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496298 Supplier Medchem Express Supplier No. HY15065
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chembridge-5861528 is a TRPA1 channel blocker that antagonizes AITC- and 4-HNE-evoked calcium influx (IC50 values are 14.3 and 18.7μM respectively).

Chembridge-5861528 is a TRPA1 channel blocker that antagonizes AITC- and 4-HNE-evoked calcium influx (IC50 values are 14.3 and 18.7μM respectively).

Specifications

Chemical Name or Material Chembridge-5861528
CAS 332117-28-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H23N5O3
Quantity 10 mg
Synonym TCS 5861528
Solubility Information DMSO : 120 mg/mL (324.83 mM; Need ultrasonic)
SMILES O=C1C2=C(N=CN2CC(NC3=CC=C(C(C)CC)C=C3)=O)N(C)C(N1C)=O
Molecular Weight (g/mol) 369.42
Formula Weight 369.42
Percent Purity 99.27%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.