accessibility menu, dialog, popup

UserName

HOOClOHCl

CAS RN 3336-41-2

IUPAC Name: 3,5-dichloro-4-hydroxybenzoic acid
Synonyms: DiClHBz
Molecular Weight (g/mol): 207.01
Molecular Formula: C7H4Cl2O3
InChi Key: AULKDLUOQCUNOK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=C(O)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results