accessibility menu, dialog, popup

UserName

CH3NOHNO(R)(S)(R)OOPOOOHONaNa

CAS RN 33430-62-5

IUPAC Name: {[3-hydroxy-5-(5-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methoxy}phosphonic acid dihydrate disodium
Synonyms: [3-hydroxy-5-(5-methyl-2,4-dioxo-3H-pyrimidin-1-yl)oxolan-2-yl]methoxyphosphonic acid dihydrate disodium
Molecular Weight (g/mol): 404.22
Molecular Formula: C10H19N2Na2O10P
InChi Key: XRJOCTOKVYURQZ-UHFFFAOYNA-N
SMILES: O.O.[Na].[Na].CC1=CN(C2CC(O)C(COP(O)(O)=O)O2)C(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)