missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymidine-5'-monophosphate disodium salt

Product Code. 15433459
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433459 1 g 1g
15453459 100 mg 100mg
15463459 250 mg 250mg
15443459 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15433459 Supplier Thermo Scientific Chemicals Supplier No. J60747.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33430-62-5 | C10H13N2Na2O8P | 366.17 g/mol

Thymidine-5'-monophosphate disodium salt is a nucleoside monophosphate used to prepare thymidine diphosphate by thymidylate kinase (TMPK) which is a nucleoside monophosphate kinase. In addition to this, it serves as a constituent of deoxyribonucleic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thymidine-5′-monophosphate disodium salt is a nucleoside monophosphate used to prepare thymidine diphosphate by thymidylate kinase (TMPK) which is a nucleoside monophosphate kinase. In addition to this, it serves as a constituent of deoxyribonucleic acid.

Solubility
Soluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33430-62-5
Molecular Formula C10H13N2Na2O8P
Molecular Weight (g/mol) 366.17
MDL Number MFCD00023797
InChI Key AGSQMPPRYZYDFV-ZJWYQBPBSA-L
Synonym tmp disodium
PubChem CID 131674985
IUPAC Name disodium;[(2R,3S,5R)-3-hydroxy-5-(5-methyl-4-oxido-2-oxopyrimidin-1-yl)oxolan-2-yl]methyl hydrogen phosphate;hydrate
SMILES [Na+].[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)NC1=O

Specifications

Color White
Quantity 1 g
Beilstein 4086927
Solubility Information Soluble in water.
Formula Weight 366.18
Physical Form Powder
Chemical Name or Material Thymidine-5'-monophosphate disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.