accessibility menu, dialog, popup

UserName

FFFFFFFFFFFFFFFF

CAS RN 335-27-3

IUPAC Name: 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis(trifluoromethyl)cyclohexane
Synonyms: perfluorodimethylcyclohexane
Molecular Weight (g/mol): 400.06
Molecular Formula: C8F16
InChi Key: LOQGSOTUHASIHI-UHFFFAOYNA-N
SMILES: FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C(F)(C(F)(F)F)C1(F)F
Boiling Point 
  • (4)
Grade 
  • (4)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)