missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(1,3-dimethylcyclohexane), tech. 80%, remainder other fluorocarbons, Thermo Scientific™

Product Code. 11377575
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377575 100 g 100g
11367575 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11377575 Supplier Thermo Scientific Alfa Aesar Supplier No. L05726.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-27-3
Molecular Formula C8F16
Molecular Weight (g/mol) 400.062
MDL Number MFCD00001469
InChI Key LOQGSOTUHASIHI-UHFFFAOYSA-N
Synonym perfluoro-1,3-dimethylcyclohexane, perfluoro 1,3-dimethylcyclohexane, 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis trifluoromethyl cyclohexane, decafluoro-1,3-bis trifluoromethyl cyclohexane, cyclohexane, perfluoro-1,3-dimethyl, cyclohexane, 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis trifluoromethyl, cyclohexane, decafluoro-1,3-bis trifluoromethyl, hexadecafluoro 1,3-dimethylcyclohexane, flutec pp3, acmc-1cjoy
PubChem CID 78975
IUPAC Name 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis(trifluoromethyl)cyclohexane
SMILES C1(C(C(C(C(C1(F)F)(F)F)(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F

Specifications

Melting Point -70°C
Density 1.828
Boiling Point 101°C to 102°C
Refractive Index 1.2908
Quantity 100 g
Beilstein 2064147
Formula Weight 400.06
Percent Purity 80%
Grade Technical
Assay remainder other fluorocarbons
Chemical Name or Material Perfluoro(1,3-dimethylcyclohexane)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.