accessibility menu, dialog, popup

UserName

CAS RN 33522-03-1

SCN

CAS RN 33522-03-1

IUPAC Name: isothiocyanatocyclopentane
Synonyms: isothiocyanatocyclopentane
Molecular Weight (g/mol): 127.21
Molecular Formula: C6H9NS
InChi Key: PJOODZCPFADLCI-UHFFFAOYSA-N
SMILES: S=C=NC1CCCC1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)