missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10582083
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10582083 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10582083 Supplier Thermo Scientific Maybridge Supplier No. TL00485DA
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 33522-03-1
CAS Min % 97.0
Density 1.03g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H9NS
Linear Formula C5H9NCS
MDL Number MFCD00040877
Quantity 1 g
Synonym cyclopentyl isothiocyanate, cyclopentane, isothiocyanato, isothiocyanato-cyclopentane, cyclopentanisothiocyanate, cyclopentylisothiocyanate, acmc-20albz
InChI Key PJOODZCPFADLCI-UHFFFAOYSA-N
SMILES C1CCC(C1)N=C=S
IUPAC Name isothiocyanatocyclopentane
Molecular Weight (g/mol) 127.205
PubChem CID 141792
Formula Weight 127.21
Percent Purity 97%
Chemical Name or Material Cyclopentyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.