accessibility menu, dialog, popup

UserName

ClONN

CAS RN 33575-83-6

IUPAC Name: 2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole
Synonyms: 2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole
Molecular Weight (g/mol): 194.62
Molecular Formula: C9H7ClN2O
InChi Key: AGLNTFQAHIRTFA-UHFFFAOYSA-N
SMILES: ClCC1=NN=C(O1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results