missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole, 97%, Thermo Scientific™

Product Code. 10678540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10678540 1 g 1g
10179819 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10678540 Supplier Thermo Scientific Acros Supplier No. 438280010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemische Identifikatoren

CAS 33575-83-6
Molecular Formula C9H7ClN2O
Molecular Weight (g/mol) 194.62
MDL Number MFCD00466332
InChI Key AGLNTFQAHIRTFA-UHFFFAOYSA-N
Synonym 2-chloromethyl-5-phenyl-1,3,4-oxadiazole, 2-chloromethyl-5-phenyl-1,3,4 oxadiazole, 5-chloromethyl-2-phenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-chloromethyl-5-phenyl, enamine_005035, chloromethylphenyloxadiazole, 1,3,4-oxadiazole,2-chloromethyl-5-phenyl, 5-chloromethyl-1,3,4-oxadiazol-2-yl benzene
PubChem CID 314941
IUPAC Name 2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole
SMILES ClCC1=NN=C(O1)C1=CC=CC=C1

Spezifikation

CAS Min % 96.0
CAS Max % 100.0
Melting Point 119.3°C to 119.7°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 194.62
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-(chloromethyl)-5-phenyl-1, 3, 4-oxadiazole
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.