accessibility menu, dialog, popup

UserName

CH3H3CCH3OOOHO

CAS RN 33693-84-4

IUPAC Name: 2-[(tert-butoxy)carbonyl]benzoic acid
Synonyms: 2-(tert-butoxycarbonyl)benzoic acid
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: PBUQZKXKYSAJDO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results