accessibility menu, dialog, popup

UserName

IH3CNH3CCH2

CAS RN 33797-51-2

IUPAC Name: (dimethylamino)methylium iodide
Synonyms: eschenmoser's salt
Molecular Weight (g/mol): 185.01
Molecular Formula: C3H8IN
InChi Key: VVDUZZGYBOWDSQ-UHFFFAOYSA-M
SMILES: [I-].CN(C)[CH2+]
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)