missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylmethyleneiminium iodide, 97%

Product Code. 10740901 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10740901 10 g Glass bottle 10g
10461051 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10740901 Supplier Thermo Scientific Acros Supplier No. 201010100
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33797-51-2 | C3H8IN | 185.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33797-51-2
Molecular Formula C3H8IN
Molecular Weight (g/mol) 185.01
MDL Number MFCD00011810
InChI Key VVDUZZGYBOWDSQ-UHFFFAOYSA-M
Synonym eschenmoser's salt, n,n-dimethylmethyleneiminium iodide, n-methyl-n-methylenemethanaminium iodide, dimethyl methylidene azanium iodide, n,n-dimethyl methyleneammonium iodide, dimethyl methylene ammonium iodide, methanaminium, n-methyl-n-methylene-, iodide, eschenmoser inverted exclamation mark s salt, eschenmoser salt, dimethyl methylene ammounium iodide
PubChem CID 2724133
IUPAC Name dimethyl(methylidene)azanium;iodide
SMILES C[N+](=C)C.[I-]

Specifications

Melting Point 219°C
Color Cream to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2C=N(CH3)2I
Quantity 10 g
Beilstein 04, IV, 153
Fieser 04,186; 08,194; 10,160; 13,121
Merck Index 15, 3275
Formula Weight 185.01
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Dimethylmethyleneiminium iodide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • N,N-Dimethylmethyleneiminium iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.