accessibility menu, dialog, popup

UserName

ClOO

CAS RN 34035-03-5

IUPAC Name: 5-(4-chlorophenyl)furan-2-carbaldehyde
Synonyms: 5-(4-chlorophenyl)furan-2-carbaldehyde
Molecular Weight (g/mol): 206.63
Molecular Formula: C11H7ClO2
InChi Key: ROJGJNINTRCMBL-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=CC=C(O1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results