missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Chlorophenyl)furfural, 95+%, Thermo Scientific™

Product Code. 10402473 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10402473 5 g Glass bottle 5g
10230443 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10402473 Supplier Thermo Scientific Acros Supplier No. 375500050
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 34035-03-5
Molecular Formula C11H7ClO2
Molecular Weight (g/mol) 206.63
MDL Number MFCD00195947
InChI Key ROJGJNINTRCMBL-UHFFFAOYSA-N
Synonym 5-4-chlorophenyl furan-2-carbaldehyde, 5-4-chlorophenyl-2-furaldehyde, 5-4-chlorophenyl furfural, 5-4-chloro-phenyl-furan-2-carbaldehyde, 2-formyl-5-4-chlorophenyl furan, 2-furancarboxaldehyde, 5-4-chlorophenyl, 5-4-chlorophenyl-2-furancarboxaldehyde, 5-4-chlorophenyl furan-2-carboxaldehyde, pubchem4001, cambridge id 7179578
PubChem CID 676184
IUPAC Name 5-(4-chlorophenyl)furan-2-carbaldehyde
SMILES ClC1=CC=C(C=C1)C1=CC=C(O1)C=O

Specifications

Melting Point 128°C to 131°C
CAS Min % 95.0
Color Brown
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Needles
Assay Percent Range 95% min. (GC)
Formula Weight 206.63
Percent Purity 95+%
Chemical Name or Material 5-(4-Chlorophenyl)furfural
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.