accessibility menu, dialog, popup

UserName

CAS RN 3405-89-8

OHOSOOCl

CAS RN 3405-89-8

IUPAC Name: 2-(4-chlorobenzenesulfonyl)acetic acid
Synonyms: (4-chlorobenzenesulfonyl)acetic acid
Molecular Weight (g/mol): 234.65
Molecular Formula: C8H7ClO4S
InChi Key: IAONCYGNBVHNCT-UHFFFAOYSA-N
SMILES: OC(=O)CS(=O)(=O)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results