CAS RN 34070-68-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Methyl-3,4-dihydro-1H-quinoxalin-2-one, TRC
CAS: 34070-68-3 Molecular Formula: C9H10N2O Molecular Weight (g/mol): 162.19 Synonym: 3,4-Dihydro-3-methyl-2(1H)-quinoxalinone,1,2-Dihydro-2-methyl-3-hydroxyquinoxaline,3,4-Dihydro-3-methyl-1H-quinoxalin-2-one,3,4-Dihydro-3-methylquinoxalin-2(1H)-one,3-Methyl-1,2,3,4-tetrahydroquinoxalin-2-one SMILES: CC1Nc2ccccc2NC1=O