CAS RN 3409-21-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[2-(Dimethylamino)ethyl]-1-indanone (>90%), TRC
CAS: 3409-21-0 Molecular Formula: C13 H17 N O Molecular Weight (g/mol): 203.2802 Synonym: 2-[2-(Dimethylamino)ethyl]-2,3-dihydro-1H-inden-1-one IUPAC Name: 2-[2-(dimethylamino)ethyl]-2,3-dihydroinden-1-one SMILES: CN(C)CCC1Cc2ccccc2C1=O