CAS RN 3413-72-7
CAS RN 3413-72-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MTIC, TRC
CAS: 3413-72-7 Molecular Formula: C5 H8 N6 O Molecular Weight (g/mol): 168.16 Synonym: 1H-Imidazole-4-carboxamide, 5-[2-(methylimino)hydrazinyl]-,1H-Imidazole-4-carboxamide, 5-(3-methyl-1-triazenyl)- (9CI),Imidazole-4(or 5)-carboxamide, 5(or 4)-(3-methyl-1-triazeno)- (7CI),Imidazole-4-carboxamide, 5-(3-methyl-1-triazeno)- (8CI),5-[2-(Methylimino)hydrazinyl]-1H-imidazole-4-carboxamide,5-(3-Methyl-1-triazeno)imidazole-4-carboxamide,5-(3-Methyltriazen-1-yl)imidazole-4-carboxamide,MTIC,NSC 407347 IUPAC Name: 5-(methylaminodiazenyl)-1H-imidazole-4-carboxamide SMILES: CNN=Nc1[nH]cnc1C(=O)N