missing translation for 'onlineSavingsMsg'
Learn More

MTIC, TRC

Product Code. 30483542
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.50mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483542 5 mg 5mg
30470171 25 mg 25mg
30467032 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483542 Supplier Toronto Research Chemicals Supplier No. TRCM7600005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3413-72-7
Molecular Formula C5 H8 N6 O
Molecular Weight (g/mol) 168.16
Synonym 1H-Imidazole-4-carboxamide, 5-[2-(methylimino)hydrazinyl]-, 1H-Imidazole-4-carboxamide, 5-(3-methyl-1-triazenyl)- (9CI), Imidazole-4(or 5)-carboxamide, 5(or 4)-(3-methyl-1-triazeno)- (7CI), Imidazole-4-carboxamide, 5-(3-methyl-1-triazeno)- (8CI), 5-[2-(Methylimino)hydrazinyl]-1H-imidazole-4-carboxamide, 5-(3-Methyl-1-triazeno)imidazole-4-carboxamide, 5-(3-Methyltriazen-1-yl)imidazole-4-carboxamide, MTIC, NSC 407347
IUPAC Name 5-(methylaminodiazenyl)-1H-imidazole-4-carboxamide
SMILES CNN=Nc1[nH]cnc1C(=O)N

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C5H8N6O/c1-7-11-10-5-3(4(6)12)8-2-9-5/h2H,1H3,(H2,6,12)(H,7,10)(H,8,9)
Formula Weight 168.076
Chemical Name or Material MTIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.