accessibility menu, dialog, popup

UserName

OHHNO

CAS RN 3416-18-0

IUPAC Name: 5-hydroxy-2,3-dihydro-1H-indol-2-one
Synonyms: 5-hydroxy-1,3-dihydroindol-2-one
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: ZGTUSQAQXWSMDW-UHFFFAOYSA-N
SMILES: OC1=CC=C2NC(=O)CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results