missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyoxindole, 96%, Thermo Scientific Chemicals

Product Code. 15668685 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15668685 5 g 5g
15658685 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15668685 Supplier Thermo Scientific Acros Supplier No. 153550050
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3416-18-0
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
InChI Key ZGTUSQAQXWSMDW-UHFFFAOYSA-N
Synonym 5-hydroxyoxindole, 5-hydroxyindolin-2-one, 5-hydroxy-1,3-dihydro-indol-2-one, 2,3-dihydro-5-hydroxyindol-2-one, 5-hydroxy-2-oxyindole, 5-hydroxy-2-indolinone, 5-hydroxy-1,3-dihydro-2h-indol-2-one, 5-hydroxy-2,3-dihydro-1h-indol-2-one, 2-indolinone, 5-hydroxy, 5-hydroxy-oxindole
PubChem CID 76955
IUPAC Name 5-hydroxy-1,3-dihydroindol-2-one
SMILES C1C2=C(C=CC(=C2)O)NC1=O

Specifications

Melting Point 266.0°C to 272.0°C
Assay Percent Range 95% min. (HPLC)
Quantity 5 g
Formula Weight 149.15
Percent Purity 96%
Chemical Name or Material 5-Hydroxyoxindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.