accessibility menu, dialog, popup

UserName

CH3OOO

CAS RN 34231-78-2

IUPAC Name: 3-formylphenyl acetate
Synonyms: 3-formylphenyl acetate
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: GVUMZPWBUAGJBP-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC(C=O)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results