CAS RN 3430-17-9
CAS RN 3430-17-9
IUPAC Name:
2-bromo-3-methylpyridine
Synonyms:
2-bromo-3-methylpyridine
Molecular Weight (g/mol):
172.03
Molecular Formula:
C6H6BrN
InChi Key:
PZSISEFPCYMBDL-UHFFFAOYSA-N
SMILES:
CC1=CC=CN=C1Br
1
–
2
of
2
results
1
2-Bromo-3-methylpyridine, 95%
CAS: 3430-17-9 Molecular Formula: C6H6BrN Molecular Weight (g/mol): 172.03 MDL Number: MFCD00239380 InChI Key: PZSISEFPCYMBDL-UHFFFAOYSA-N Synonym: 2-bromo-3-picoline,2-bromo-3-methylpyridin,2-bromo-3-methyl pyridine,2-bromo-3-methyl-pyridine,3-methyl-2-bromopyridine,pyridine, 2-bromo-3-methyl,2-brom-3-methylpyridin,pubchem9208,2-bromo-3-mehtylpyridine PubChem CID: 220832 IUPAC Name: 2-bromo-3-methylpyridine SMILES: CC1=CC=CN=C1Br