missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylpyridine, 98+%

Product Code. 15499089
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499089 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15499089 Supplier Thermo Scientific Alfa Aesar Supplier No. L19522.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3430-17-9 | C6H6BrN | 172.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3430-17-9
Density 1.544
Boiling Point 218°C to 219°C
Flash Point >110°C (230°F)
Molecular Formula C6H6BrN
Refractive Index 1.5695
MDL Number MFCD00239380
Quantity 5 g
Beilstein 107896
Synonym 2-bromo-3-picoline, 2-bromo-3-methylpyridin, 2-bromo-3-methyl pyridine, 2-bromo-3-methyl-pyridine, 3-methyl-2-bromopyridine, pyridine, 2-bromo-3-methyl, 2-brom-3-methylpyridin, pubchem9208, 2-bromo-3-mehtylpyridine
InChI Key PZSISEFPCYMBDL-UHFFFAOYSA-N
SMILES CC1=CC=CN=C1Br
IUPAC Name 2-bromo-3-methylpyridine
Molecular Weight (g/mol) 172.03
PubChem CID 220832
Formula Weight 172.03
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-3-methylpyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.