accessibility menu, dialog, popup

UserName

CAS RN 34413-35-9

NN

CAS RN 34413-35-9

IUPAC Name: 5,6,7,8-tetrahydroquinoxaline
Synonyms: 5,6,7,8-tetrahydroquinoxaline
Molecular Weight (g/mol): 134.18
Molecular Formula: C8H10N2
InChi Key: XCZPDOCRSYZOBI-UHFFFAOYSA-N
SMILES: C1CCC2=NC=CN=C2C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results