missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydroquinoxaline, 99%

Product Code. 11387626
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11387626

Brand: Thermo Scientific Alfa Aesar L11204.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34413-35-9 | C8H10N2 | 134.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34413-35-9
Molecular Formula C8H10N2
Molecular Weight (g/mol) 134.182
MDL Number MFCD00038078
InChI Key XCZPDOCRSYZOBI-UHFFFAOYSA-N
Synonym cyclohexapyrazine, quinoxaline, 5,6,7,8-tetrahydro, unii-m153cj9ra0, fema no. 3321, 5.6.7.8-tetrahydroquinoxaline, nutty quinoxaline, pubchem15475, acmc-209i7e, dsstox_cid_27678, dsstox_rid_82495
PubChem CID 36822
IUPAC Name 5,6,7,8-tetrahydroquinoxaline
SMILES C1CCC2=NC=CN=C2C1

Specifications

Density 1.061
Boiling Point 72°C to 74°C (4 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.542
Quantity 5 g
Beilstein 957053
Formula Weight 134.18
Percent Purity 99%
Chemical Name or Material 5,6,7,8-Tetrahydroquinoxaline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.