accessibility menu, dialog, popup

UserName

CAS RN 345-83-5

IUPAC Name: (4-fluorophenyl)(phenyl)methanone
Synonyms: (4-fluorophenyl)(phenyl)methanone
Molecular Weight (g/mol): 200.21
Molecular Formula: C13H9FO
InChi Key: OGTSHGYHILFRHD-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)