accessibility menu, dialog, popup

UserName

OOO

CAS RN 3459-92-5

IUPAC Name: dibenzyl carbonate
Synonyms: dibenzyl carbonate
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: PIZLBWGMERQCOC-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)OCC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results