Learn More
Dibenzyl carbonate, 98%
Descrizione
Ionic liquids can effectively accelerate slow N-benzylation reactions utilizing dibenzyl carbonate as an alkylating reagent. Dibenzyl carbonate (DBzlC) has been used to benzylate phenylacetonitrile, benzyl phenylacetate and phenol. Selective N, N-dibenzylation of primary aliphatic amines was carried with dibenzyl carbonate in the presence of phosphonium salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquids can effectively accelerate slow N-benzylation reactions utilizing dibenzyl carbonate as an alkylating reagent. Dibenzyl carbonate (DBzlC) has been used to benzylate phenylacetonitrile, benzyl phenylacetate and phenol. Selective N, N-dibenzylation of primary aliphatic amines was carried with dibenzyl carbonate in the presence of phosphonium salts.
Solubility
Sparingly Soluble (0.099 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifica
Specifica
| Melting Point | 27°C to 30°C |
| Boiling Point | 150°C to 155°C (0.8 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.5485 |
| Quantity | 250 g |
| Beilstein | 1882864 |
| Sensitivity | Air sensitive |
| Solubility Information | Sparingly Soluble (0.099g/L) (25°C). |
| Formula Weight | 242.27 |
| Percent Purity | 98% |
| Vedi altri risultati |
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.