accessibility menu, dialog, popup

UserName

H3N(S)

CAS RN 34698-41-4

IUPAC Name: (1S)-2,3-dihydro-1H-inden-1-aminium
Synonyms: (1S)-2,3-dihydro-1H-inden-1-aminium
Molecular Weight (g/mol): 134.2
Molecular Formula: C9H12N
InChi Key: XJEVHMGJSYVQBQ-VIFPVBQESA-O
SMILES: [NH3+][C@H]1CCC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)