accessibility menu, dialog, popup

UserName

NH2F

CAS RN 348-54-9

IUPAC Name: 2-fluoroaniline
Synonyms: 2-fluoroaniline
Molecular Weight (g/mol): 111.12
Molecular Formula: C6H6FN
InChi Key: FTZQXOJYPFINKJ-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)