accessibility menu, dialog, popup

UserName

CH3ONHH2N

CAS RN 34801-09-7

IUPAC Name: N-(2-aminophenyl)acetamide
Synonyms: N-(2-aminophenyl)acetamide
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: MPXAYYWSDIKNTP-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results