accessibility menu, dialog, popup

UserName

CAS RN 34803-66-2

IUPAC Name: 1-(pyridin-2-yl)piperazine
Synonyms: pyridinylpiperazine
Molecular Weight (g/mol): 163.22
Molecular Formula: C9H13N3
InChi Key: GZRKXKUVVPSREJ-UHFFFAOYSA-N
SMILES: C1CN(CCN1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)