missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyridyl)piperazine, 99%

Product Code. 11317455
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317455 25 g 25g
11307455 5 g 5g
11327455 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11317455 Supplier Thermo Scientific Alfa Aesar Supplier No. L05451.14
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34803-66-2 | C9H13N3 | 163.224 g/mol

1-(2-Pyridyl)piperazine may be employed as reagent for the determination of both aliphatic and aromatic isocyanates in air by reversed-phase HPLC. It may be employed as reagent for the fluorometric determination of airborne diisocyantes. It is a compound that belongs to a class of selective α2-adrenoceptor antagonists. 1-(2-Pyridyl)piperazine shows sympatholytic activity. 1-(2-Pyridyl)piperazine is also a metabolite of Azaperone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Pyridyl)piperazine may be employed as reagent for the determination of both aliphatic and aromatic isocyanates in air by reversed-phase HPLC. It may be employed as reagent for the fluorometric determination of airborne diisocyantes. It is a compound that belongs to a class of selective α2-adrenoceptor antagonists. 1-(2-Pyridyl)piperazine shows sympatholytic activity. 1-(2-Pyridyl)piperazine is also a metabolite of Azaperone.

Solubility
Soluble in Chloroform, Methanol.

Notes
Air Sensitive. Store away from air and oxidizing agents. Incompatible with carbondioxide, Strong acids, Acid chlorides, Acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34803-66-2
Molecular Formula C9H13N3
Molecular Weight (g/mol) 163.224
MDL Number MFCD00006216
InChI Key GZRKXKUVVPSREJ-UHFFFAOYSA-N
Synonym 1-2-pyridyl piperazine, 1-pyridin-2-yl piperazine, 1-pyridin-2-yl-piperazine, 1-2-pyridinyl piperazine, piperazine, 1-2-pyridinyl, 2-pyridylpiperazine, pyridinylpiperazine, 2-1-piperazinyl pyridine
PubChem CID 94459
IUPAC Name 1-pyridin-2-ylpiperazine
SMILES C1CN(CCN1)C2=CC=CC=N2

Specifications

Density 1.072
Boiling Point 120°C to 122°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.595
Quantity 25 g
UN Number UN2735
Beilstein 140423
Sensitivity Air Sensitive
Solubility Information Soluble in Chloroform,Methanol.
Formula Weight 163.22
Percent Purity 99%
Chemical Name or Material 1-(2-Pyridyl)piperazine
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2-Pyridyl)piperazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1-(2-Pyridyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.