accessibility menu, dialog, popup

UserName

NNHNN

CAS RN 34803-68-4

IUPAC Name: 2-(piperazin-1-yl)pyrazine
Synonyms: 2-(piperazin-1-yl)pyrazine
Molecular Weight (g/mol): 164.21
Molecular Formula: C8H12N4
InChi Key: HCGFLVDMFDHYJD-UHFFFAOYSA-N
SMILES: C1CN(CCN1)C1=CN=CC=N1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results