missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyrazinyl)piperazine, 98%

Product Code. 10456252 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10456252 1 g Glass bottle 1g
10068971 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10456252 Supplier Thermo Scientific Acros Supplier No. 367130010
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 34803-68-4 | C8H12N4 | 164.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34803-68-4
Molecular Formula C8H12N4
Molecular Weight (g/mol) 164.21
MDL Number MFCD00040766
InChI Key HCGFLVDMFDHYJD-UHFFFAOYSA-N
Synonym 2-piperazin-1-yl pyrazine, 1-2-pyrazinyl-piperazine, 1-piperazinylpyrazine, 1-pyrazin-2-yl piperazine, 1-2-pyrazinyl piperazine, pyrazin-2-yl-piperazine, 3,4,5,6-tetrahydro-2h-1,2' bipyrazinyl, 2-piperazinylpyrazine, 2-piperazinopyrazine, 1-piperazinyl pyrazine
PubChem CID 2734639
IUPAC Name 2-piperazin-1-ylpyrazine
SMILES C1CN(CCN1)C2=NC=CN=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Boiling Point 105°C to 108°C (0.5 Mbar)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 164.21
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1-(2-Pyrazinyl)piperazine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2-Pyrazinyl)piperazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.