accessibility menu, dialog, popup

UserName

CAS RN 35094-87-2

IUPAC Name: 2,4,5-trihydroxybenzaldehyde
Synonyms: 2,4,5-trihydroxybenzaldehyde
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H6O4
InChi Key: WNCNWLVQSHZVKV-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C(C=O)C=C1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results