accessibility menu, dialog, popup

UserName

CAS RN 351-36-0

CH3OHNFFF

CAS RN 351-36-0

IUPAC Name: N-[3-(trifluoromethyl)phenyl]acetamide
Synonyms: M-(trifluorome)acetanilide
Molecular Weight (g/mol): 203.16
Molecular Formula: C9H8F3NO
InChi Key: HNIPNANLYHXYDE-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results