accessibility menu, dialog, popup

UserName

FFFBrSOONH2

CAS RN 351003-63-9

IUPAC Name: 2-bromo-4-(trifluoromethyl)benzene-1-sulfonamide
Synonyms: 2-bromo-4-(trifluoromethyl)benzenesulfonamide
Molecular Weight (g/mol): 304.08
Molecular Formula: C7H5BrF3NO2S
InChi Key: IJUIBMWZUXWVLI-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=C(Br)C=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results