accessibility menu, dialog, popup

UserName

BaOSOOS

CAS RN 35112-53-9

IUPAC Name: barium(2+) 1-oxo-1λ⁶-disulfene-1,1-bis(olate)
Synonyms: barium(2+) Hypo
Molecular Weight (g/mol): 249.44
Molecular Formula: BaO3S2
InChi Key: ONPIOWQPHWNPOQ-UHFFFAOYSA-L
SMILES: [Ba++].[O-]S([O-])(=O)=S
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)