missing translation for 'onlineSavingsMsg'
Learn More

Barium thiosulfate, pure, Thermo Scientific™

Product Code. 15239856 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15239856 500 g 500g
15229856 100 g 100g
15219856 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15239856 Supplier Thermo Scientific Acros Supplier No. 453815000
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 35112-53-9
Molecular Formula BaO3S2
Molecular Weight (g/mol) 249.46
InChI Key ONPIOWQPHWNPOQ-UHFFFAOYSA-L
Synonym barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo
PubChem CID 169663
IUPAC Name barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane
SMILES [O-]S(=O)(=S)[O-].[Ba+2]

Specifications

Quantity 500 g
Linear Formula BaS2O3
Merck Index 15,995
Solubility Information Solubility in water: slightly soluble. Other solubilities: insoluble in alcohol
Formula Weight 249.46
Percent Purity 97.5 to 102.5%
Grade Pure
Chemical Name or Material Barium thiosulfate
compliance-icons
Product Identifier
  • Barium thiosulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.