accessibility menu, dialog, popup

UserName

FI

CAS RN 352-34-1

IUPAC Name: 1-fluoro-4-iodobenzene
Synonyms: benzene, 1-fluoro-4-iodo-
Molecular Weight (g/mol): 222
Molecular Formula: C6H4FI
InChi Key: KGNQDBQYEBMPFZ-UHFFFAOYSA-N
SMILES: FC1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (3)