accessibility menu, dialog, popup

UserName

CH3F

CAS RN 352-70-5

IUPAC Name: 1-fluoro-3-methylbenzene
Synonyms: M-fluorotoluene
Molecular Weight (g/mol): 110.13
Molecular Formula: C7H7F
InChi Key: BTQZKHUEUDPRST-UHFFFAOYSA-N
SMILES: CC1=CC=CC(F)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results