accessibility menu, dialog, popup

UserName

H3COO

CAS RN 3524-62-7

IUPAC Name: 2-methoxy-1,2-diphenylethan-1-one
Synonyms: 2-methoxy-1,2-diphenylethanone
Molecular Weight (g/mol): 226.28
Molecular Formula: C15H14O2
InChi Key: BQZJOQXSCSZQPS-UHFFFAOYNA-N
SMILES: COC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results